This product will no longer be available on thermofisher.com after Dec. 28, 2020. Learn more »
Thermo Scientific™
Superfrost Plus Gold Control Adhesion Slide 3/4 in. White Tab
Related applications:
Anatomical Pathology
Simplify staining and record keeping with Thermo Scientific Superfrost Plus Histology Control Adhesion Slides. Place required controls and patient tissue on the same slide. The control section is easily identified with a bright red, etched or gold box. Superfrost Plus Histology Control Adhesion Slides hold tissue sections on permanently without the need for expensive coatings in IHC and Anatomical Pathology applications. Control slides for use in automated instrumentation have a control box on the back surface of the slide. Available in several different combinations. For use in histology, cytology, and microbiology applications.
Back to top
Features:
  • Smooth marking area for effortless writing, automated printing, or printed labels
  • Manufactured from pure, water-white glass
  • Precleaned for immediate use
  • Item ES-3012PLUS Features Superfrost Plus Gold Adhesion Coating
  • Item ES-325 is a Non-Adhesion Slide