Image-iT™ Cell Painting Kit, Sufficient for ten 384- or 96-well plates - FAQs
Image-iT™ Cell Painting Kit, Sufficient for ten 384- or 96-well plates - FAQs
View additional product information for Image-iT™ Cell Painting Kit - FAQs (I65000, I65500)
1 product FAQs found
How many plates can I stain using the Image-iT Cell Painting Kit (Cat. No. I65500)?
The Image-iT Cell Painting Kit (Cat. No. I65500) contains sufficient reagents to stain 10 x 96‑well plates.