iGlow™ Blue Sample Tray - FAQs
iGlow™ Blue Sample Tray - FAQs
View additional product information for iGlow™ Blue Sample Tray - FAQs (IGA012)
7 product FAQs found
What nucleic acid gel stains are supported by the iGlow Gel Documentation System?
The iGlow Gel Documentation System supports a wide variety of nucleic acid stains, including ethidium bromide, SYBR Safe, SYBR Gold, SYBR Green, GelRed, and RedSafe stains.
What maintenance is required for the iGlow Gel Documentation System and imaging accessories?
No routine calibration or replacement parts are required. When needed, the sample trays, touchscreen, and transilluminator glass may be cleaned using deionized water, 70% ethanol, or isopropyl alcohol followed by wiping with a lint-free cloth or tissue.
Is there a performance advantage to using either the blue or UV LEDs on the iGlow Gel Documentation System?
Blue LED illumination offers a safer alternative to UV light and does not cause DNA degradation. Most fluorescent DNA and protein stains are compatible with blue illumination, although some stains, such as ethidium bromide, may provide stronger signal intensity with UV illumination.
How are sample trays installed in the iGlow Gel Documentation System?
Each sample tray includes notches that align with locating pegs on the sample drawer. After installation, the iGlow Gel Documentation System automatically recognizes the inserted tray and activates the corresponding workflow.
What accessories are included with the iGlow Gel Documentation System?
The iGlow Gel Documentation System includes the UV, blue, and white trays required for imaging, the UV Safety Guard for gel excision, and the sample tray holder.
How does the iGlow Gel Documentation System automatically recognize which sample tray is inserted?
The iGlow Gel Documentation System uses integrated sensors to detect when a sample tray is inserted and automatically enables the corresponding workflow on the user interface.
Are the iGlow imaging trays compatible with other instrumentation?
No. The imaging trays are designed specifically for use with the iGlow Gel Documentation System.