What protein stains are supported by the iGlow Gel Documentation System?
The iGlow Gel Documentation System supports a wide variety of protein stains, including Coomassie-based stains, Pierce Silver Stain, Coomassie Fluor Orange stain, Flamingo stain, and SYPRO stains, including Ruby, Red, Orange, and Tangerine stains.
What maintenance is required for the iGlow Gel Documentation System and imaging accessories?
No routine calibration or replacement parts are required. When needed, the sample trays, touchscreen, and transilluminator glass may be cleaned using deionized water, 70% ethanol, or isopropyl alcohol followed by wiping with a lint-free cloth or tissue.
How are sample trays installed in the iGlow Gel Documentation System?
Each sample tray includes notches that align with locating pegs on the sample drawer. After installation, the iGlow Gel Documentation System automatically recognizes the inserted tray and activates the corresponding workflow.
What accessories are included with the iGlow Gel Documentation System?
The iGlow Gel Documentation System includes the UV, blue, and white trays required for imaging, the UV Safety Guard for gel excision, and the sample tray holder.
How does the iGlow Gel Documentation System automatically recognize which sample tray is inserted?
The iGlow Gel Documentation System uses integrated sensors to detect when a sample tray is inserted and automatically enables the corresponding workflow on the user interface.
Are the iGlow imaging trays compatible with other instrumentation?
No. The imaging trays are designed specifically for use with the iGlow Gel Documentation System.