Nautilus LIMS™ for Dynamic Discovery and R&D Environments
Thermo Scientific™
Nautilus LIMS™ for Dynamic Discovery and R&D Environments
Need a LIMS that can keep up with your dynamic laboratory? This flexible, configurable system increases efficiency, throughput and data reliability while simplifying compliance.
Have Questions?
Catalog NumberType
INF-12000Nautilus LIMS
Catalog number INF-12000
Price (USD)
-
Type:
Nautilus LIMS
To accelerate new discoveries and get products to market as quickly as possible, R&D and manufacturing labs have to reconfigure and change on the fly. Data management can't be a bottleneck.
Developed in partnership with customers in fast-paced R&D environments, the Thermo Scientific™ Nautilus LIMS™ for Dynamic Discovery and R&D Environments is a highly flexible, easily configurable system that increases workflow efficiency, throughput and data reliability while simplifying administration, sample traceability and regulatory compliance.
  • Easy-to-use toolkits allow quick configuration to keep up with the pace of operations, helping managers make better-informed decisions, faster than ever
  • Automated complex plate-handling functionality and patented graphical utilities facilitate data tracking and management; even nonexpert users can map and track processes with ease
  • Clients can create workflows, plot sample lifecycles and automate actions across disparate platforms while still integrating regulated processes that adhere to GLP and 21 CFR Part 11 guidelines
  • A flexible, intuitive interface—fully accessible using a Windows™ Client or standard web service—graphically maps laboratory workflows to meet the needs of even the most rapidly changing environments
Increases Throughput and Productivity
  • Simplifies the definition of plate configurations and fill patterns, letting users build and format work action and decision-tree processes against project requirements
  • Enables increased throughput, advanced plate handling and automation via intuitive graphical workflows, plate editors and built-in integration functionality
  • Improves both productivity and accuracy of results by eliminating manual, error-prone processes
  • Integrates LIMS and analytical systems within a single interface, allowing users to directly compare analyses from disparate instruments
Reduces Total Cost of Ownership
  • Built-in instrument integration and full plate-handling functionality delivers significant productivity gains right out of the box
  • Standard integration functionality allows data to be easily imported—without customized coding—from a variety of analytical instruments
  • Instrument integration enables rapid access to up-to-the-minute raw and derived data that impact every phase of laboratory processes
Fully Supports Regulatory Compliance
  • Database design utilizes an Oracle™ RDBMS to enforce data security, auditability and compliance; unique system architecture enables integration with the entire laboratory in a flexible, secure manner that ensures the data authenticity, integrity and traceability
  • System further supports 21 CFR Part 11 compliance through the use of configurable signature templates and advanced audit capabilities
  • An intuitive web interface facilitates communication and interaction with systems throughout the organization as well as with partner facilities and regulatory agencies
Specifications
DescriptionNautilus
For Use With (Equipment)Nautilus
TypeNautilus LIMS
Unit SizeEach