Product Image
Invitrogen™

Enhancer Solution

The InstantOne ELISA Enhancer Solution is a supplement added to the 5X Cell Lysis Buffer (Cat. No. IOCLB1) to yieldRead more
Have Questions?
Catalog NumberQuantity
IOES1100 mL
Catalog number IOES1
Price (USD)
152.65
Online Exclusive
157.00
Save 4.35 (3%)
Each
Quantity:
100 mL
Price (USD)
152.65
Online Exclusive
157.00
Save 4.35 (3%)
Each
The InstantOne ELISA Enhancer Solution is a supplement added to the 5X Cell Lysis Buffer (Cat. No. IOCLB1) to yield the Cell Lysis Mix solution that is applied to cultured cells or tissues prior to running the assays. The 1X Cell Lysis Mix is prepared by simply diluting Cell Lysis Mix (5X) (a mixture of the Cell Lysis Buffer (5X) and the Enhancer Solution) 5-fold with deionized water. This buffer is used to lyse cells after the removal of culture medium, and is typically used to lyse adherent cells or non-adherent cells that have been harvested by centrifugation. Lysis Mix should be used as the diluent for any dilution of cellular lysates that are required.

Reported Application
ELISA

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)ELISA
FormLiquid
Quantity100 mL
Reagent TypeSignal Enchancers
Storage Requirements2°C to 8°C
Validated ApplicationELISA
Unit SizeEach