Product Image
Invitrogen™

InstantOne ELISA Wash Buffer (10X)

The InstantOne ELISA Wash Buffer (10X) is developed for use with the InstantOne ELISA Cell Signaling assays. The InstantOne ELISARead more
Have Questions?
Catalog NumberQuantity
IOWB18 mL
Catalog number IOWB1
Price (MXN)
-
Quantity:
8 mL
The InstantOne ELISA Wash Buffer (10X) is developed for use with the InstantOne ELISA Cell Signaling assays. The InstantOne ELISA Wash Buffer is supplied as a 10X concentrate and is used for washing the InstantOne ELISA cell signaling assay microplate. It is a simple mix of buffers, salts, and mild detergent. If washing wells with a microplate washer, use 3x washes with a 10 second mixing cycle.
The InstantOne ELISA Wash Buffer (10X) is included with every InstantOne ELISA sold. This wash buffer is offered additionally as a stand-alone item in cases where experimental procedures and/or automation require much higher volumes of wash buffer.

Reported Application
ELISA

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialWash Buffer
For Use With (Application)ELISA
For Use With (Equipment)InstantOne ELISA Cell Signaling assays
Physical FormLiquid
Quantity8 mL
Unit SizeEach