Vanquish™ Flex UHPLC Systems
Vanquish™ Flex UHPLC Systems
Vanquish™ Flex UHPLC Systems
Vanquish™ Flex UHPLC Systems
Vanquish™ Flex UHPLC Systems
Thermo Scientific™

Vanquish™ Flex UHPLC Systems

Green features
Discover new benchmarks in accuracy, precision and sensitivity with Thermo Scientific™ Vanquish™ Flex UHPLC systems. Vanquish Horizon UHPLC Systems are an ACT labeled product with a My Green Lab ACT Ecolabel.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
IQLAAAGABHFAPUMBJCVanquish Flex Binary UHPLC System
IQLAAAGABHFAPUMBHVVanquish™ Flex Quaternary UHPLC System
Catalog number IQLAAAGABHFAPUMBJC
Price (USD)
-
Type:
Vanquish Flex Binary UHPLC System

Obtain new benchmarks in accuracy, precision and sensitivity with the Thermo Scientific™ Vanquish™ Flex UHPLC systems. Providing biocompatibility with a state-of-the-art quaternary or binary high-pressure solvent blending, these ultra-high performance liquid chromatography systems share all Vanquish values, such as a design focused on uptime, robustness and reliability. Multiple detection options give you the performance and flexibility you need. These system are ideal for method development or high-throughput analyses and can be seamlessly coupled to a mass spectrometer (MS).

View Vanquish Flex UHPLC resources ∣ Product tour ∣ Build your HPLC ∣ HPLC Support Center

Drives the separation with more flexibility

Extend your capabilities by adding all the flexibility you would expect from leading-edge isocratic, low-pressure quaternary, or high-pressure binary solvent blending. Our SmartFlow pumping technology with automatic compensation for changing eluent compressibility ensures excellent flow and gradient precision, independent of eluent composition and backpressures. This gives you unrivaled data confidence and flexibility in method transfer.

  • 1000 bar (15,000 psi) of pump pressure and flow rates up to 8 mL/min
  • Industry-leading flow and gradient precision
  • 4 solvent channels for quaternary solvent mixing; up to 6 solvent channels for binary high-pressure solvent mixing

Handles the sample with more accuracy

Out of the box, Vanquish Flex Split Sampler FT module comes with a vial capacity of 216 samples. Increase to 8832 sample capacity (23 well plates) by adding the optional Vanquish Charger module.

  • SmartInject technology and unsurpassed sample dosage precision
  • Innovative air stream cooling for maximum sample integrity in both the sampler and the charger module
  • Automated workflows with barcode reading

Controls the separation with more confidence

Ensure even temperature throughout using the two thermostatting modes of the Vanquish Column Compartment H module. Avoid performance loss from temperature effects using the system's active pre-heating to ensure solvent entering the column will match the column temperature.

  • Two thermostatting modes: still air and forced air
  • Temperature range from 5°C to 80°C
  • Active pre-heating and post column cooling
  • Extendable with valve switching options

Detects the analyte with more versatility

Ensure you collect the most information possible from your separation, and benefit from multiple detection options for the Vanquish platform so that no peak remains unseen.

  • Highly dependable diode array detection
  • Highly selective and sensitive fluorescence detection
  • Near universal response for label-free detection of non-chromophoric substances with charged aerosol detection

Driven by gold-standard CDS Software

The Vanquish Flex system seamlessly integrates with our award winning Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS). Designed with Operational Simplicity™ in mind, the Chromeleon CDS realizes the promise of the Vanquish platform—easier interaction delivering a new UHPLC experience.

Easy and reliable tool-free connections

Thermo Scientific™ Viper™ fingertight fittings have revolutionized the way HPLC connections are made. The Vanquish Flex system utilizes Viper fittings in all connections from the pump to the detector. Whether setting up a method or maintaining the system, the Viper fingertight fitting system allows any user to make consistently perfect connections.

Sustainability

The Vanquish Horizon UHPLC Systems are an ACT labeled product with a My Green Lab ACT Ecolabel.

The My Green Lab ACT Ecolabel serves as the premier ecolabel for lab products, encompassing consumables, chemicals, and equipment. Standing for Accountability, Consistency, and Transparency (ACT), this certification offers clear, standardized information about a product's environmental footprint across its life cycle covering manufacturing, energy and water use, packaging, and end-of-life disposal. Leverage the My Green Lab ACT Ecolabel to make informed purchasing choices based on transparent environmental data so you can advance sustainability without sacrificing performance. Learn more about the ACT Ecolabel and participating Thermo Scientific products here.

Specifications
For Use With (Equipment)2 channel binary high-pressure gradient
Green FeaturesACT Ecolabel (Platinum)
Sample Temperature Range4°C to 40°C in 0.1°C increments
TypeVanquish Flex Binary UHPLC System
Column Temperature Range5°C to 120°C in 0.1°C increments (18°C below ambient)
Detector Data Collection Rate250 Hz Max.
Max. Pressure100 MPa
Solvent Channels2
Unit SizeEach

Frequently asked questions (FAQs)

What maintains temperature control and method reproducibility in the Vanquish Flex UHPLC System?

Column Compartment H provides still air/forced air modes, active preheating, and broad temperature ranges for both the column and samples, supporting consistent retention and peak shape.

How many samples can we queue on a Vanquish Flex UHPLC System? Can it run all weekend?

The autosampler holds 4 sample plates by default and can scale to 8,832 samples (23 plates) with the Vanquish Sample Hub module for extended unattended operation.

What are the performance limits of a Vanquish Flex UHPLC System?

The Vanquish Flex UHPLC System delivers up to 1000 bar (15,000 psi) and 8 mL/min, with industry-leading flow/gradient precision via SmartFlow technology.

Should I choose a binary or quaternary Vanquish Flex UHPLC System for method development and routine runs?

Choose quaternary for straightforward low-pressure mixing across 4 solvent channels; choose binary for high-pressure mixing of two solvents out of up to 6 channels when you want fast, precise gradients and solvent flexibility.

What’s special about the Vanquish Neo UHPLC System’s autosampler design for proteomics?

The split-loop autosampler minimizes sample dispersion, provides thorough washing of fluidics and columns, and decreases maintenance while enabling fast loading/equilibration—ideal for high-sensitivity and high-throughput methods.