Determine blending endpoints and ingredient concentrations in real time using the Thermo Scientific™ Antaris™ Target Blend Analyzer. This compact, powerful NIR was developed in partnership with pharmaceutical industry leaders and provides continuity and scalability between product development and operations, while also facilitating the implementation of control strategies. The Antaris Target Blend Analyzer is the most complete, reliable analytical package for monitoring solids blending, including traceable design modeled from quality system requirements in the highly-regulated medical device industry.
Error loading your content!
Catalog number | Price ({{currency}}) | Availability | Qty | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
{{product.sku}}
also known as {{product.formattedSku}} |
—
Add To Cart
Save to list
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Antaris Target Blend Analyzer Features:
Ordering Information:
The Antaris Target Blend Analyzer is sold as a configured system designed to meet your specific application needs. For more information, contact your Thermo Scientific Sales Representative.