DELTA™ Q Isotope Ratio Mass Spectrometer
Thermo Scientific™
DELTA™ Q Isotope Ratio Mass Spectrometer
Perform all isotope ratio mass spectrometry (IRMS) applications using a single analyzer with DELTA Q IRMS systems
Have Questions?
Catalog NumberType
IQLAAEGAATFABHMZZZIRMS
Catalog number IQLAAEGAATFABHMZZZ
Price (EUR)
-
Type:
IRMS
The Thermo Scientific™ DELTA™ Q Isotope Ratio Mass Spectrometer (IRMS) represents the next evolutionary leap forwards in isotope analysis, combining unprecedented performance with an easy-to-use innovative Thermo Scientific™ Qtegra™ Intelligent Scientific Data Solution (ISDS) Software. The DELTA Q IRMS is designed to be seamlessly connected with a wide range of Thermo Scientific™ peripherals, aimed at supporting varying investigations including food authenticity, criminal and environmental forensics, doping control, and scientific research.
Outstanding sensitivity
  • Sensitivities up to 800 molecules per CO2 ion in dual-inlet mode and 1,000 molecules per ion in continuous-flow mode.
Versatility to tackle diverse applications
  • Up to 10 collectors in a wide range of configurations, supporting all isotope ratio MS applications in the mass range up to m/z 96.
  • Simultaneous mass range up to ±25%.
  • Available preconcentrators, elemental analyzers, GC interfaces, LC interfaces, and continuous flow inlets.
  • On-line H2 capability.
Highly robust and reliable design
  • Monolithic analyzer with fixed alignment of all ion optical components, including the electromagnet, ensures unprecedented reliability and robustness.
  • Integrated signal amplifiers and digitizers eliminate connections for greater reliability.
  • Fixed-position ion optics require no alignment during installation or maintenance.
  • Optimized pumping strategy enables single-stage turbo pumping for all normal continuous-flow applications.
  • Comprehensive, automated diagnostics driven by Qtegra ISDS Software are provided for performance analysis.
Compact and user-friendly design
  • Small footprint requires less lab space.
  • Space for online peripheral placed on top.
  • Dual inlet, μ-volume and multiport are inside and out of the way.
  • Continuous flow interfaces can be controlled from the front panel.
  • All pumps are located internally for less clutter and noise.
Qtegra ISDS Software
  • Dramatically improves your lab productivity with automatic Get Ready functionality, ready-to-go templates and inbuilt quality control tools.
  • Carefully optimized intelligent workflows for gas IRMS drive you from sample to result, eliminating any barriers and flexibly switching between different system configurations.
  • The Qtegra ISDS Software platform provides a wide range of features, which ensure the utmost integrity and traceability for all of your raw data, metadata, and reported results.
Specifications, terms, and pricing are subject to change. Not all products are available in all countries. Please consult your local sales representative for details.
Specifications
TypeIRMS
Unit SizeEach
Frequently asked questions (FAQs)
How user-friendly is the DELTA Q Isotope Ratio Mass Spectrometer system in terms of software, automation, and maintenance?
The Thermo Scientific DELTA Q IRMS uses the Thermo Scientific Qtegra ISDS Software platform, offering automatic Get Ready functionality, workflow automation (sample to result), ready-to-go templates, inbuilt quality-control tools, and full data/metadata traceability.
On the hardware side, the analyzer is monolithic with fixed alignment of ion-optical components, requiring no manual intervention for day-to-day operation. The ion source is easily accessible for routine maintenance.
How does the DELTA Q Isotope Ratio Mass Spectrometer handle challenging sample types, low sample amounts, or diverse inlets?
The Thermo Scientific DELTA Q Isotope Ratio Mass Spectrometer system supports continuous-flow as well as dual-inlet modes, enabling flexibility for bulk gas samples or small volume samples.
The high sensitivity and excellent signal-to-noise performance help when dealing with low-level signals or small sample amounts.
Its versatile inlet connectivity (GC, LC, elemental analyzers, universal on-line gas preparation and introduction systems) helps handle complex sample matrices, and allows isotope analysis at bulk or compound-specific level.
What kinds of sample types does the DELTA Q Isotope Ratio Mass Spectrometer measure?
The Thermo Scientific DELTA Q Isotope Ratio Mass Spectrometer supports measurement of stable isotopes across a wide range of sample types, such as food, environmental and petrochemical samples, forensics samples, geological materials, and gases.
What isotope-ratio capabilities does the DELTA Q Isotope Ratio Mass Spectrometer support?
The Thermo Scientific DELTA Q Isotope Ratio Mass Spectrometer supports measurement of stable isotopes such as C, N, S, O, and H across a wide range of sample types using the Dual Inlet system and changeover valve or by Continuous Flow gas introduction via dedicated peripherals.