FastQuan™ System for High-Throughput LC-MS
FastQuan™ System for High-Throughput LC-MS
FastQuan™ System for High-Throughput LC-MS
Thermo Scientific™
FastQuan™ System for High-Throughput LC-MS
Delivering fast, robust screening and confirmation results without compromising data quality.
Have Questions?
Catalog NumberFor Use With (Equipment)Includes
IQLAAEGAAXFAOTMBHZLC-MS SystemUser Guide, 3-Day LSMS Training Course at Customer Site (North America Only)
Catalog number IQLAAEGAAXFAOTMBHZ
Price (MXN)
-
For Use With (Equipment):
LC-MS System
Includes:
User Guide, 3-Day LSMS Training Course at Customer Site (North America Only)
Increase laboratory productivity with an affordable, robust, high-throughput LC-MS system bundle. Today's clinical research and forensic toxicology laboratories demand more productivity, reliability, and cost-efficiency even when analyzing tough biological matrices.
The industry-proven Thermo Scientific™ FastQuan™ system bundle delivers fast, robust, economical screening of analytes, and confirmation results without compromising quality of data. The value-priced, FastQuan system bundle is built on twenty years of multichannel innovation with the Thermo Scientific™ Transcend™ II LX-2 system combined with the Thermo Scientific™ TSQ Endura™ triple quadrupole mass spectrometer and Thermo Scientific™ TraceFinder™ software.
Increase productivity and reduce cost per analysis
The FastQuan system bundle brings the productivity of two reproducible channels of UHPLC separation to one mass spectrometer. As analytes elute for only a portion of a UHPLC run, mass spectrometers can be efficiently utilized to collect data by reducing mass spectrometer idle time and enabling mass spectrometers to achieve nearly 100% utilization. The FastQuan solution improves clinical research and forensic toxicology laboratory throughput and operational cost while streamlining your LC-MS workflows, resulting in lower cost per sample analysis. Optional upgrades to four-channel operation or online sample cleanup further increase your productivity.
Acquire, process and analyze data, with ease and confidence
The FastQuan system bundle includes TraceFinder software, an easy-to-use workflow-driven solution for clinical research and forensic labs performing targeted screening of analytes and quantitative confirmatory methods using LC-MS. The software is designed for easy implementation of the requirements in clinical research and forensic labs. Simplified data acquisition, powerful data processing options, batch analysis, comprehensive data review, and extensive reporting features increase productivity.
Maximize day-to-day productivity
As the natures of the matrices and molecules you must analyze evolve, so does the need to develop robust, sensitive methods to address molecules of all types. Our application support scientists are available to train users and help improve your method development processes, enabling your lab to achieve organizational profitability goals.
To maximize productivity and uptime from day one, the FastQuan high-throughput system bundle includes three days of onsite training (North America only) and a three-year end-to-end maintenance package.
Specifications
TypeSystem Bundle
Mass Spectrometer InterfaceThermo Scientific TSQ Endura MS - Including Easy Max NG Source, Syringe, 6 Port Valve, TSQ Quantiva/Endura Pre-Installation Kit
Liquid Chromatography SystemTranscend II, LX-2 W/UltiMate Pumps
SoftwareThermo Scientific TraceFinder 3.3 SP1
For Use With (Equipment)LC-MS System
IncludesUser Guide, 3-Day LSMS Training Course at Customer Site (North America Only)
Unit SizeEach