ARL™ SMS-2300 Robotics-Based Automation for OES or XRF
Actual product may vary
ARL™ SMS-2300 Robotics-Based Automation for OES or XRF
ARL™ SMS-2300 Robotics-Based Automation for OES or XRF
ARL™ SMS-2300 Robotics-Based Automation for OES or XRF
Meet stringent steel specifications while increasing efficiency and lowering production costs. The Thermo Scientific™ ARL™ SMS-2300 Robotics-Based Automation for OES or XRF is an automated quality control solution to optimize your steel production operation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
IQLAAHGABDFAAZMBJIOES Robotics-based Automation
IQLAAHGABMFAAZMBJJXRF Robotics-based Automation
Catalog number IQLAAHGABDFAAZMBJI
Price (BRL)
-
Type:
OES Robotics-based Automation
With the high-performance ARL SMS-2300 system, even the most complex samples are prepared and analyzed under reproducible conditions with reduced operator interaction, so human variables and subjective factors are eliminated. The result is increased output, reliable results and reduced costs.
The ARL SMS-2300 Automation for OES or XRF delivers powerful, flexible automation designed to reduce routine operator interaction with Thermo Scientific™ ARL™ 4460, ARL™ iSpark, and ARL™ iSpark Plus Optical Emission Spectrometers, as well as ARL™ 9900 and ARL™ X900 X-Ray Fluorescence (XRF) Spectrometers.
Main Features
  • Designed for easy integration into an automated workflow / automated laboratory integration.
  • The full system can be mounted in the Thermo Scientific™ ARL™ QuantoShelter™ System and placed very close to the production process.
  • Tasks are performed in parallel; samples are prepared while other samples are introduced or analyzed; the instrument stand cleaning is performed in hidden time by the spectrometer, independently of the ARL SMS-2300 robot.
  • More samples processed with the addition of automatic sample preparation solutions including the Haas Mini Mill with Thermo Scientific™ ARL™ PrepControl™ System and Thermo Scientific™ ARL™ EasyPrep™ Software.
  • Direct access to the Haas Mini Mill milling machine to reduce processing times for ferrous samples. Direct access to other preparation machines also possible (To be defined during project layout).
  • Factory configured and customized.
  • Compact chassis and the simplest installation ever.
  • Integrated electronics.
  • Better analysis dependability and quality.
  • Very rapid payback.
  • Faster results availability.
  • Reduces routine operator tasks.
  • Manufactured and maintained according to ISO 9001 procedures.
Advanced Technology
  • Innovative Fanuc™ Industrial Robot
  • Thermo Scientific™ OXSAS™ Analytical Software
Specifications
TypeOES Robotics-based Automation
Unit SizeEach