Can the Thermo Scientific ARL X'TRA Companion XRD be used for routine quality control?
Yes. The Thermo Scientific ARL X'TRA Companion XRD is designed for routine quality assurance and quality control (QA/QC) environments where fast, reliable phase analysis is essential. Automated workflows, intuitive software, and optional sample changers help laboratories achieve consistent workflows with minimal operator training.
What applications is the Thermo Scientific ARL X'TRA Companion XRD optimized for?
The Thermo Scientific ARL X'TRA Companion XRD is optimized for phase identification, quantitative phase analysis, mineralogical analysis, crystallinity determination, and quality control. Common applications include cement manufacturing, mining and geology, battery materials, ceramics, pharmaceuticals, advanced materials, environmental research, and academic laboratories.
How does the Thermo Scientific ARL X'TRA Companion XRD improve laboratory productivity?
The Thermo Scientific ARL X'TRA Companion XRD is designed to improve laboratory productivity through automated sample handling, rapid data collection, integrated phase identification, one-click Rietveld refinement, and streamlined reporting. These features reduce manual intervention, improve workflow consistency, and enable faster turnaround for routine analyses.
What is crystallinity in XRD?
Crystallinity describes how ordered the atoms are within a material's crystal structure. XRD measures crystallinity by analyzing diffraction patterns, helping researchers evaluate crystal quality, compare processing conditions, monitor material performance, and characterize materials such as polymers, pharmaceuticals, and battery materials.
Can XRD measure amorphous materials?
Yes. While XRD is primarily used to identify and quantify crystalline phases, it can also estimate the amorphous content of a material when used with appropriate quantitative analysis methods, such as Rietveld refinement or PONCKS with an internal standard. This capability is important for applications including cement, glass, ceramics, pharmaceuticals, and advanced materials research.
Can XRD identify unknown materials?
Yes. X-ray diffraction is one of the most effective techniques for identifying unknown crystalline materials. By comparing a sample's diffraction pattern with reference databases, XRD can identify individual crystalline phases and compounds, making it valuable for failure analysis, contamination investigations, quality control, and research applications.
When should I use XRD instead of XRF?
Use XRD when you need to identify crystalline phases and use XRF when you need rapid elemental analysis to determine which elements are present and at what concentrations. Together, XRD and XRF provide complementary information that supports quality control, process optimization, and materials characterization.
What is the difference between XRD and XRF?
Although both techniques use X-rays, they answer different analytical questions.
- XRD identifies crystalline phases and crystal structures, revealing what compounds or minerals are present.
- XRF measures elemental composition, determining which elements are present and in what concentrations.
Many laboratories use both techniques because they answer different analytical questions.