When is XRD useful for quality control in cement and mining workflows?
XRD is useful when crystalline phase information is needed to complement elemental analysis techniques such as XRF. In cement production, phase information can help monitor clinker mineralogy and support process control. In mining and minerals processing, mineralogical information can help characterize ores and support processing decisions.
What is the difference between the Thermo Scientific ARL X'TRA Pro XRD and the Thermo Scientific ARL X'TRA Companion XRD?
The Thermo Scientific ARL X'TRA Companion X-ray Diffractomber is a benchtop system designed for routine XRD analysis, while the Thermo Scientific ARL X'TRA Pro X-ray Diffractometer is designed for laboratories requiring higher sample throughput, greater automation, and increased analytical capacity for demanding industrial QA/QC workflows. Consider the ARL X'TRA Pro XRD when throughput and automation are important workflow requirements.
Can the Thermo Scientific ARL X'TRA Pro XRD be integrated with XRF workflows?
Yes. The Thermo Scientific ARL X'TRA Pro X-ray Diffractometer is designed to support integration with laboratory automation and complementary analytical technologies through an OEM automation protocol. This capability can support workflows incorporating XRD and XRF (X-ray fluorescence) when both phase and elemental information are required.
How does the Thermo Scientific ARL X'TRA Pro XRD support automated data analysis and reporting?
The Thermo Scientific ARL X'TRA Pro X-ray Diffractometer is designed to streamline the workflow from data acquisition through results with automated data analysis and reporting capabilities. LIMS connectivity can also help integrate XRD results into laboratory data-management workflows.
How many samples can the Thermo Scientific ARL X'TRA Pro XRD handle?
The Thermo Scientific ARL X'TRA Pro X-ray Diffractometer automated XY sample handling accommodates up to 98 samples. The system is designed to support throughput of 50–100+ samples per day, helping laboratories manage higher sample volumes with less routine operator intervention.
Does the Thermo Scientific ARL X'TRA Pro XRD automate sample handling?
Yes. The Thermo Scientific ARL X'TRA Pro XRD uses automated XYZ sample handling to help reduce repetitive manual sample-loading tasks and support high-throughput laboratory workflows.
How does the Thermo Scientific ARL X'TRA Pro XRD support high-throughput XRD workflows?
The Thermo Scientific ARL X'TRA Pro X-ray Diffractometer combines automated sample handling with a high-power X-ray source and streamlined data acquisition and analysis. Together, these capabilities are designed to support higher sample volumes with less routine operator intervention.
What applications does Thermo Scientific ARL X'TRA Pro XRD support?
The Thermo Scientific ARL X'TRA Pro X-ray Diffractometer is designed primarily for high-throughput QA/QC applications in the cement and mining industries. XRD phase analysis can support applications such as clinker phase monitoring in cement production and mineralogical characterization in mining and minerals processing.
When should I choose the Thermo Scientific ARL X'TRA Pro XRD for my laboratory?
The Thermo Scientific ARL X'TRA Pro X-ray Diffractometer is designed for laboratories that require high-throughput X-ray diffraction analysis for industrial QA/QC workflows. It combines automated sample handling with XRD data acquisition and analysis capabilities to support routine mineralogical and phase analysis.