What advantages does WDXRF provide for complex sample matrices?
WDXRF provides higher spectral resolution than EDXRF, helping reduce peak overlaps and improve elemental separation in complex sample matrices. This can support more accurate quantification when analyzing samples containing multiple elements with overlapping spectral characteristics.
What factors should I consider when selecting a WDXRF spectrometer?
When selecting a WDXRF spectrometer, laboratories typically evaluate sample types, required detection limits, elemental coverage, throughput requirements, automation needs, software capabilities, regulatory requirements, and long-term analytical objectives. Matching instrument capabilities to application requirements can help optimize laboratory performance and workflow efficiency.
Can the ARL PERFORM'X support both routine quality control and advanced research workflows?
Yes. The Thermo Scientific ARL PERFORM'X Sequential X-Ray Fluorescence (WDXRF) Spectrometer is designed to support routine production testing as well as advanced research applications. Its combination of broad elemental coverage, flexible analytical capabilities, and quantitative performance allows laboratories to use a single system across multiple workflows.
What capabilities make the ARL PERFORM'X suitable for trace elemental analysis?
The Thermo Scientific ARL PERFORM'X Sequential X-Ray Fluorescence (WDXRF) Spectrometer combines high spectral resolution, excellent precision, broad dynamic range, and low detection limits to support demanding trace elemental analysis applications. These capabilities make it suitable for quality control, regulatory compliance, raw material characterization, environmental analysis, and advanced research workflows.
Which laboratories are best suited for the ARL PERFORM'X WDXRF Spectrometer?
The Thermo Scientific ARL PERFORM'X Sequential X-Ray Fluorescence (WDXRF) Spectrometer is well suited for laboratories that require high-precision elemental analysis, including mining, metals, cement, petrochemical, environmental, geological, materials science, and industrial quality control laboratories.
When should I choose the ARL PERFORM'X instead of an EDXRF system?
The Thermo Scientific ARL PERFORM'X Sequential X-Ray Fluorescence (WDXRF) Spectrometer is often selected when applications require higher spectral resolution, improved peak separation, enhanced quantitative performance, or lower detection limits than those typically achieved with EDXRF systems.
What is the difference between WDXRF and EDXRF?
Wavelength Dispersive X-ray Fluorescence (WDXRF) uses diffraction crystals to separate X-ray wavelengths, providing higher spectral resolution and improved peak separation than EDXRF. WDXRF is commonly selected for applications requiring enhanced quantitative performance, lower detection limits, and analysis of complex sample matrices.
Can the ARL PERFORM'X WDXRF Spectrometer analyze unknown materials without calibration standards?
Yes. The Thermo Scientific ARL PERFORM'X Sequential X-Ray Fluorescence (WDXRF) Spectrometer supports standardless analysis through Thermo Scientific UniQuant and QuantAS software packages. These tools can help users identify and quantify the elemental composition of unknown materials when calibration standards are unavailable.
What sample types can the ARL PERFORM'X WDXRF Spectrometer analyze?
The Thermo Scientific ARL PERFORM'X Sequential X-Ray Fluorescence (WDXRF) Spectrometer can analyze a wide range of solid and liquid sample types, including ores, metals, alloys, cement, glass, ceramics, refractories, polymers, fuels, lubricants, chemicals, environmental samples, and geological materials.
What is the ARL PERFORM'X WDXRF Spectrometer used for?
The Thermo Scientific ARL PERFORM'X Sequential X-Ray Fluorescence (WDXRF) Spectrometer is used for advanced elemental analysis of solids and liquids. It supports major, minor, and trace element analysis across industries including mining, metals, cement, petroleum, chemicals, polymers, glass, refractories, environmental research, geochemistry, and materials science.