253 Plus 10 kV Isotope Ratio MS
253 Plus 10 kV Isotope Ratio MS
253 Plus 10 kV Isotope Ratio MS
Thermo Scientific™
253 Plus 10 kV Isotope Ratio MS
Utilize the new gold standard in IRMS. The new Thermo Scientific™ 253 Plus™ builds on the robustness, linearity, and sensitivity only available in the Thermo Scientific 10 kV IRMS technology.
Have Questions?
Número do catálogoType
IQLAAMGAATFADEMBHWIRMS
Número do catálogo IQLAAMGAATFADEMBHW
Preço (BRL)
-
Type:
IRMS
Utilize the gold standard in IRMS. The Thermo Scientific™ 253 Plus™ 10 kV IRMS redefines high performance for isotope ratio mass spectrometry. It builds on the robustness, linearity, and sensitivity only available in the Thermo Scientific 10 kV IRMS technology. Technical innovations for the 253 Plus 10 kV IRMS improve sample utilization, stability, background and signal-to-noise resulting in ultimate precision on smallest sample sizes.
Proven Thermo Scientific 10 kV IRMS technology
  • “Extra bright” 10 kV EI ion source enables highest sensitivity and linearity in IRMS
  • All-metal sealed ion source and analyzer with gold gaskets
  • The only IRMS with variable conductance (VISC)
  • Analytical dynamic range greater than 107
  • Hydrogen continuous flow capability
  • Fully integrated workflow solutions in Thermo Scientific™ Qtegra™ ISDS Software delivering easy measurements setup, fully automated operation and streamlined data evaluation 
Especificações
TypeIRMS
Unit SizeEach