Savant™ ISS110 SpeedVac™ Kit
Thermo Scientific™  Related applications: Vacuum Concentrators & Pumps
Process large batches of DNA/RNA samples efficiently with the easy-to-use Thermo Scientific™ Savant™ ISS110 SpeedVac™ Kit. An ideal choice, these medium capacity kits are complete systems, containing everything you need under one part number.
Error loading your content!
  Catalog number Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
You saved {{product.yourPrice.saved}}
{{product.availability.message}} Add To Cart
Showing {{showMoreLimit}} of {{commerceData.products.length}} |  Showing {{shownProducts.length}} of {{commerceData.products.length}} | 
Back to top
Description
Features:
  • Built-in ETFE coated vacuum pump provides years of lasting service
  • 4 liters, -50°C refrigerated vapor trap integrated
  • Choice of three drying rates
Solvents and Solvent Combinations:
  • PCR buffers (aqueous)
  • Ethanol and water
  • Isopropanol
Includes:
  • Basic integrated concentrator
  • 64× 1.5mL Rotor
  • CryoCool Heat Transfer Fluid (1L)
Recommended for:
  • DNA/RNA preparation prior to analysis by electrophoresis or microarrays
  • Labeling cDNA with fluorescent dyes
  • Drying PCR samples in aqueous buffers
Featured brochure
  Savant™ SpeedVac™ Concentrators and SpeedVac Kits brochure ›
Rotor selection guide
  Rotors and Carriers Ordering Information ›
Documents
Manuals & protocols