Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Fisher Chemical™

Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™

ClH, CAS Number-7647-01-0, chloorwaterstof, anhydrous hydrochloric acid, acide chlorhydrique, hydrogen chloride hcl, hydrogen chloride, chlorohydric acid, chlorwasserstoff, hydrochloric acid, spirits of salt, muriatic acid, 1L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol | CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
J-4320-15
also known as J/4320/15
1 LHDPE plastic bottle
Catalog number J-4320-15
also known as J/4320/15
Price (USD)
-
Quantity:
1 L
Packaging:
HDPE plastic bottle
ClH, CAS Number-7647-01-0, chloorwaterstof, anhydrous hydrochloric acid, acide chlorhydrique, hydrogen chloride hcl, hydrogen chloride, chlorohydric acid, chlorwasserstoff, hydrochloric acid, spirits of salt, muriatic acid, 1L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol
Ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP
Specifications
Boiling Point100°C
CAS7647-01-0
ChEBICHEBI:17883
ColorColorless
Formula Weight36.46g/mol
IUPAC Namechlorane
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
MDL NumberMFCD00011324
Molecular FormulaClH
PackagingHDPE plastic bottle
Physical FormLiquid
PubChem CID313
SMILESCl
Synonymhydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Molecular Weight (g/mol)36.46
Quantity1 L
Unit SizeEach