Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Fisher Chemical™

Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
5 L
Packaging:
View all
Plastic container
Catalog number J-4320-21
also known as J/4320/21
Price (TWD)
-
Quantity:
5 L
Packaging:
Plastic container
Request bulk or custom format
Specifications
Boiling Point100°C
CAS7647-01-0
ChEBICHEBI:17883
ColorColorless
Formula Weight36.46g/mol
View more
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis