Hydrochloric Acid Solution 1M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Hydrochloric Acid Solution 1M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Hydrochloric Acid Solution 1M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Fisher Chemical™

Hydrochloric Acid Solution 1M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™

ClH, CAS Number-7647-01-0, chloorwaterstof, anhydrous hydrochloric acid, hydrogen chloride hcl, chlorohydric acid, muriatic acid, spirits of salt, chlorwasserstoff, hydrochloric acid, acide chlorhydrique, hydrogen chloride, 6AMP, 57 deg.C, CHEBI:17883, Colorless, 36.45g/mol | CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
J-4320C-90
also known as J/4320C/90
6 AmpulesAmpoule
Catalog number J-4320C-90
also known as J/4320C/90
Price (USD)
-
Quantity:
6 Ampules
Packaging:
Ampoule
ClH, CAS Number-7647-01-0, chloorwaterstof, anhydrous hydrochloric acid, hydrogen chloride hcl, chlorohydric acid, muriatic acid, spirits of salt, chlorwasserstoff, hydrochloric acid, acide chlorhydrique, hydrogen chloride, 6AMP, 57 deg.C, CHEBI:17883, Colorless, 36.45g/mol
Specifications
Boiling Point57°C
CAS7647-01-0
ChEBICHEBI:17883
ColorColorless
Formula Weight36.45g/mol
IUPAC Namechlorane
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
MDL NumberMFCD00011324 MFCD00792839
Melting Point-35°C
Molecular FormulaClH
PackagingAmpoule
Physical FormLiquid
PubChem CID313
SMILESCl
Synonymhydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Vapor Pressure125mbar at 20°C
Viscosity1.9 mPaS at 15°C
Molecular Weight (g/mol)36.46
Quantity6 Ampules
pH1
Unit SizeEach