Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Fisher Chemical™

Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Have Questions?
產品號碼QuantityPackaging
J-7950-21
亦稱為 J/7950/21
5 LHDPE plastic bottle
產品號碼 J-7950-21
亦稱為 J/7950/21
價格 (TWD)
-
Quantity:
5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
規格
CAS7772-98-7
ChEBICHEBI:32150
IUPAC Namedisodium sulfanidesulfonate
InChI KeyAKHNMLFCWUSKQB-UHFFFAOYSA-L
MDL NumberMFCD00003499
檢視更多
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis