HEPES, 1M Solution, pH 7.3, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
HEPES, 1M Solution, pH 7.3, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
500 mL
Packaging:
Nalgene bottle
Catalog number J16924.K2
Price (USD)/ Each
242.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
285.00 
Save 42.35 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
Nalgene bottle
Request bulk or custom format
Price (USD)/ Each
242.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
285.00 
Save 42.35 (15%)
Add to cart
HEPES, 1M Solution, pH 7.3, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Catalog numberJ16924.K2
Price (USD)/ Each
242.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
285.00 
Save 42.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7365-45-9
IUPAC Name2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular FormulaC8H18N2O4S
InChI KeyJKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILESOCCN1CCN(CCS(O)(=O)=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
DNase(endo): Passes test
Protease activityPasses test
DNase(exo): Passes test
RNasePasses test
RUO – Research Use Only