You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Buffers and Standards
▸
Life Science Buffers
▸
HEPES, ≥99% (Dry Basis), Ultrapure
Element Search
Structure Search
Thermo Scientific Chemicals
HEPES, ≥99% (Dry Basis), Ultrapure
CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Have Questions?
Change view
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Try the chem dex
TM
web app
Catalog number
J16926.A1
Price (EUR)
411,65
Online Exclusive
457,00
Save 45,35 (10%)
Each
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
411,65
Online Exclusive
457,00
Save 45,35 (10%)
Each
Add to cart
Chemical Identifiers
CAS
7365-45-9
IUPAC Name
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular Formula
C8H18N2O4S
InChI Key
JKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILES
OCCN1CCN(CCS(O)(=O)=O)CC1
View more
Specifications
Specification Sheet
Loss on Drying
≤ 0.5%
Identification (FTIR)
Conforms
Assay (unspecified)
Absorbance (250 nm, 1M, 1 cm): ≤ 0.2
Trace Metal
Heavy Metals(Pb): ≤ 10 ppm
Assay (unspecified)
≥ 99.0% (dry basis)
View more
RUO – Research Use Only