Paraformaldehyde Solution, 4% in PBS
Thermo Scientific Chemicals

Paraformaldehyde Solution, 4% in PBS

 
Have Questions?
Catalog NumberQuantity
J19943.K21 L
Catalog number J19943.K2
Price (USD)
85.65
Special offer
Online exclusive
Ends: 28-Dec-2026
101.00
Save 15.35
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
85.65
Special offer
Online exclusive
Ends: 28-Dec-2026
101.00
Save 15.35
Each
Specifications
Chemical Name or MaterialParaformaldehyde Solution
Concentration4% in PBS
Health Hazard 1H317-H318-H351
Health Hazard 3P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c
MDL NumberMFCD00133991
View more
 
RUO – Research Use Only

Frequently asked questions (FAQs)

Does 4% Paraformaldehyde in PBS contain methanol as a stabilizer?

No, 4% Paraformaldehyde in PBS does not contain methanol as a stabilizer.

At what temperature do you recommend storing 4% Paraformaldehyde in PBS (Cat. Nos. J61899.xx and J19943.K2)?

We recommend storing 4% Paraformaldehyde in PBS (Cat. Nos. J61899.xx and J19943.K2) at 2-8 degrees C.