Thermo Scientific Chemicals

Sodium Chloride, ACS, ≥99%, Ultrapure, Thermo Scientific Chemicals

Catalog number: J21618.A9
10 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium Chloride, ACS, ≥99%, Ultrapure, Thermo Scientific Chemicals

Catalog number: J21618.A9
10 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: J21618.A9
Price (USD)
Price: 229.00
Online price: 194.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7647-14-5
IUPAC Name
sodium chloride
Molecular Formula
ClNa
InChI Key
FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES
[Na+].[Cl-]
Form
White/clear crystalline powder
Assay (unspecified)
≥ 99.0%
Residual halide
Iodide (I): ≤ 0.002%
Impurity content
Phosphate (PO₄): ≤ 5 ppm
Trace Metal
Iron (Fe): ≤ 2 ppm

Description

Used for the precipitation of DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text