Yeast Extract, Ultrapure
Thermo Scientific Chemicals

Yeast Extract, Ultrapure

 
Have Questions?
Catalog NumberQuantity
J23547.A11 kg
Catalog number J23547.A1
Price (EUR)
79,20
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
79,20
Each
Chemical Identifiers
CAS8013-01-2
SpecificationsSpecification SheetSpecification Sheet
CommentSource: BakerÆs Yeast
Assay (unspecified)Protein (TN x 6.25): 57-72%
Assay (unspecified)Salmonella: Negative
Assay (unspecified)Standard Plate Count: ≤ 15,000/gm
Residual water≤ 6% at time of packaging
View more
 
  • Format: Off-white powder
  • CAS: 8013-01-2
  • MDL Number: MFCD00132599
RUO – Research Use Only