Thermo Scientific Chemicals

Naloxone hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: J60013.MC
100 mg, Each
Thermo Scientific Chemicals

Naloxone hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: J60013.MC
100 mg, Each
Quantity
Catalog number: J60013.MC
also known as J60013-MC
Price (USD)

Chemical Identifiers

CAS
357-08-4
IUPAC Name
hydrogen (1S,5R,13R,17S)-10,17-dihydroxy-4-(prop-2-en-1-yl)-12-oxa-4-azapentacyclo[9.6.1.0¹,¹³.0⁵,¹⁷.0⁷,¹⁸]octadeca-7,9,11(18)-trien-14-one chloride
Molecular Formula
C19H22ClNO4
InChI Key
RGPDIGOSVORSAK-STHHAXOLSA-N
SMILES
[H+].[Cl-].OC1=CC=C2C[C@H]3N(CC=C)CC[C@@]45[C@@H](OC1=C24)C(=O)CC[C@@]35O
Appearance (Color)
White to pale cream
Assay from Suppliers CofA
≥97.5% (dry wt.)
Loss on Drying
≤11.0%
Optical Rotation
-170.5° ±5.5°
Form
Crystals or powder or crystalline powder

Description

A specific opiate antagonist. Blocks the action of sigma-agonists at opioid sites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Naloxone hydrochloride the hydrochloride salt of the anti-opioid naloxone
Application
• Naloxone is a fast-acting inverse agonist of the mu-opioid receptor that can be used to competitively inhibit the binding of opioid compounds
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text