Thiamine nitrate
Thiamine nitrate
Thiamine nitrate
Thermo Scientific Chemicals

Thiamine nitrate

CAS: 532-43-4 | C12H17N5O4S | 327.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Catalog number J60014.09
also known as J60014-09
Price (EUR)
43,40
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
43,40
Each
Chemical Identifiers
CAS532-43-4
IUPAC Name3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-3H-1λ⁴,3-thiazol-1-ylium nitrate
Molecular FormulaC12H17N5O4S
InChI KeyUIERGBJEBXXIGO-UHFFFAOYSA-N
SMILES[O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98% min.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only