Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane hydrochloride, 1M soln., pH 8.0, RNase free

Catalog number: J60080.AK
250 mL, Each
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane hydrochloride, 1M soln., pH 8.0, RNase free

Catalog number: J60080.AK
250 mL, Each
Quantity
Catalog number: J60080.AK
also known as J60080-AK
Price (USD)
Price: 78.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
1185-53-1
IUPAC Name
hydrogen 2-amino-2-(hydroxymethyl)propane-1,3-diol chloride
Molecular Formula
C4H12ClNO3
InChI Key
QKNYBSVHEMOAJP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC(CO)(CO)CO
pH
8.0 ?0.1
Comment
Prepared in RNase free molecular biology grade water and filtered through a 0.22micron filter.
Concentration
1M Tris hydrochloride
Appearance (Color)
Colorless
Form
Liquid

Description

Tris is used in pH control in buffering systems for electrophoresis applications.Tris is used in assays used to characterize the activity and kinetics of the enzymes that catalyze SUMOylation of Small ubiquitin-like proteins (SUMO) and SUMO-dependent protein-protein interactions. Also used as buffers in a wide variety of biological systems. It has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text