Glycine, 0.2M buffer soln., pH 3.5
Glycine, 0.2M buffer soln., pH 3.5
Glycine, 0.2M buffer soln., pH 3.5
Thermo Scientific Chemicals

Glycine, 0.2M buffer soln., pH 3.5

CAS: 56-40-675.067 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J60154.AK
also known as J60154-AK
Price (EUR)
66,00
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
66,00
Each
Specifications
CAS56-40-6
OdorOdorless
Recommended StorageAmbient Temperatures
Concentration0.2M buffer soln.
Molecular Weight (g/mol)75.067
View more
Glycine, is used as a compound for polyacrylamide gel electrophoresis and Western blots. It is also used widely as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers (Tris-Glycine; sc-296648) where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. It is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine, is used as a compound for polyacrylamide gel electrophoresis and Western blots. It is also used widely as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers (Tris-Glycine; sc-296648) where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. It is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

Solubility
It is soluble in water (100 mg/ml), and acetone (slightly soluble). Insoluble in alcohol, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kuhse et al. The inhibitory glycine receptor: architecture, synaptic localization and molecular pathology of a postsynaptic ion-channel complex. Curr.Opin.Neurobiol. 1995, 3,(6), 338-44.
  2. Betz. Glycine receptors: heterogeneous and widespread in the mammalian brain. Trends Neurosci. 1991, 14,(10), 458-61.