Thermo Scientific Chemicals

N-Decanoyl-N-methyl-D-glucamine, Thermo Scientific Chemicals

Catalog number: J60173.03
1 g, Each
Thermo Scientific Chemicals

N-Decanoyl-N-methyl-D-glucamine, Thermo Scientific Chemicals

Catalog number: J60173.03
1 g, Each
Quantity
Catalog number: J60173.03
also known as J60173-03
Price (USD)
Price: 108.00
Online price: 92.65
Your price:
Quantity
-

Chemical Identifiers

CAS
85261-20-7
IUPAC Name
N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]decanamide
Molecular Formula
C17H35NO6
InChI Key
UMWKZHPREXJQGR-XOSAIJSUSA-N
SMILES
CCCCCCCCCC(=O)N(C)C[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
Appearance (Color)
White
Form
Powder
Solution Test
50mg/ml in methanol
Conductivity
40?mhos max., 0.3% w/v in water at 25?C

Description

Non-ionic detergent for solubilizing membrane proteins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text