Thermo Scientific Chemicals

Oxonic acid potassium salt, Thermo Scientific Chemicals

Catalog number: J60400.14
25 g, Each
Thermo Scientific Chemicals

Oxonic acid potassium salt, Thermo Scientific Chemicals

Catalog number: J60400.14
25 g, Each
Quantity
Catalog number: J60400.14
also known as J60400-14
Price (USD)
Price: 136.00
Online price: 116.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2207-75-2
IUPAC Name
potassium 4,6-dioxo-1,4,5,6-tetrahydro-1,3,5-triazine-2-carboxylate
Molecular Formula
C4H2KN3O4
InChI Key
IAPCTXZQXAVYNG-UHFFFAOYSA-M
SMILES
[K+].[O-]C(=O)C1=NC(=O)NC(=O)N1
Form
Powder
Appearance (Color)
White to off-white
Assay from Supplier's CofA
≥97.0%
Infrared spectrum
Conforms

Description

Oxonic acid potassium salt is an inhibitor of uricase. This product has been used for the inhibition of 5-fluorouracil-induced gastrointestinal toxicity without the loss of its antitumor activity in rats. It has also been used to induce hyperuricemia in rats; as it inhibits uric acid metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxonic acid potassium salt is an inhibitor of uricase. This product has been used for the inhibition of 5-fluorouracil-induced gastrointestinal toxicity without the loss of its antitumor activity in rats. It has also been used to induce hyperuricemia in rats; as it inhibits uric acid metabolism.

Solubility
Soluble in hot water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text