HEPES, 1.0M buffer soln., pH 9.0
HEPES, 1.0M buffer soln., pH 9.0
HEPES, 1.0M buffer soln., pH 9.0
Thermo Scientific Chemicals

HEPES, 1.0M buffer soln., pH 9.0

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J60440.AP
also known as J60440-AP
Price (TWD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
CAS7365-45-9
ChEBICHEBI:42334
Chemical Name or MaterialHEPES buffer soln.
IUPAC Name2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
InChI KeyJKMHFZQWWAIEOD-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only