Thermo Scientific Chemicals

Chlorogenic acid, Thermo Scientific Chemicals

Catalog number: J60457.MD
250 mg, Each
Thermo Scientific Chemicals

Chlorogenic acid, Thermo Scientific Chemicals

Catalog number: J60457.MD
250 mg, Each
Quantity
Catalog number: J60457.MD
also known as J60457-MD
Price (USD)
Quantity
-

Chemical Identifiers

CAS
327-97-9
IUPAC Name
(1S,3R,4R,5R)-3-{[(2E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy}-1,4,5-trihydroxycyclohexane-1-carboxylic acid
Molecular Formula
C16H18O9
InChI Key
CWVRJTMFETXNAD-JUHZACGLSA-N
SMILES
O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O
Assay (Titration)
95.0% min.

Description

Chlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.

Solubility
Soluble in ethanol and acetone.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text