Thermo Scientific Chemicals

Buspirone hydrochloride, Thermo Scientific Chemicals

Catalog number: J60476.06
5 g, Each
Thermo Scientific Chemicals

Buspirone hydrochloride, Thermo Scientific Chemicals

Catalog number: J60476.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60476.06
also known as J60476-06
Price (USD)
Price: 222.00
Online price: 191.65
Your price:
Quantity
-

Description

It is widely used in cell signaling and neuroscience research. Buspirone has been used to mitigate the electrically stimulated 5-hydroxytryptamine outflow from rat cortical slices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is widely used in cell signaling and neuroscience research. Buspirone has been used to mitigate the electrically stimulated 5-hydroxytryptamine outflow from rat cortical slices.

Solubility
Soluble in methanol (50 mg/ml), water (partly), DMSO (100 mM).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text