BICINE, 0.5M buffer soln.,  pH 7.5
BICINE, 0.5M buffer soln.,  pH 7.5
BICINE, 0.5M buffer soln.,  pH 7.5
Thermo Scientific Chemicals

BICINE, 0.5M buffer soln., pH 7.5

CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J60494.AK
also known as J60494-AK
Price (TWD)
-
Quantity:
250 mL
Request bulk or custom format
Specifications
CAS150-25-4
ChEBICHEBI:40957
Chemical Name or MaterialBICINE buffer soln.
IUPAC Name2-[bis(2-hydroxyethyl)amino]acetic acid
InChI KeyMFBDBXAVPLFMNJ-UHFFFAOYSA-M
View more
A zwitterionic amino acid buffer agent used for low temperature work. The use of bicine in a thin layer ion exchange chromatography method for protein resolution has been published. Bicine has been utilized in peptide and protein crystallization. A kinetic study of a quaternary transition-state analogue complex of creatine kinase used bicine in the reaction buffer. A multiphasic buffer system for SDS-PAGE of proteins and peptides that incorporates bicine has been described.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A zwitterionic amino acid buffer agent used for low temperature work. The use of bicine in a thin layer ion exchange chromatography method for protein resolution has been published. Bicine has been utilized in peptide and protein crystallization. A kinetic study of a quaternary transition-state analogue complex of creatine kinase used bicine in the reaction buffer. A multiphasic buffer system for SDS-PAGE of proteins and peptides that incorporates bicine has been described.

Solubility
Soluble in water (1M) at 20°C, ethanol (slightly), ether (slightly), and chloroform (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. P Walde.; R Wick.; M Fresta.; A Mangone. Autopoietic self-reproduction of fatty acid vesicles. J. Am. Chem. Soc. 1994, 116, (26), 11649-11654.
  2. SS Mansy.; JP Schrum.; M Krishnamurthy.; S Tob. Template-directed synthesis of a genetic polymer in a model protocell. Nature. 2008, 454, 122-125.