Manidipine dihydrochloride
Manidipine dihydrochloride
Manidipine dihydrochloride
Manidipine dihydrochloride
Thermo Scientific Chemicals

Manidipine dihydrochloride

Calcium channel blocker | CAS: 89226-75-5 | C35H40Cl2N4O6 | 683.627 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60496.MB
also known as J60496-MB
Price (TWD)
-
Request A Quote
Quantity:
25 mg
Request bulk or custom format
Specifications
CAS89226-75-5
Chemical Name or MaterialManidipine dihydrochloride
ColorYellow
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Formula Weight683.63
View more
Calcium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only