SSC (20X), RNAse free
Thermo Scientific Chemicals

SSC (20X), RNAse free

Have Questions?
變更視圖buttonViewtableView
Quantity:
250 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 J60561.AK
亦稱為 J60561-AK
價格 (TWD)
-
Quantity:
250 mL
Request bulk or custom format
規格
Chemical Name or MaterialSSC
Recommended StorageAmbient temperatures
Concentration20X
Physical FormLiquid
Quantity250 mL
檢視更多
SSC solution is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in Southern and Northern blotting, dot blotting, and colony and plaque lifts. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
SSC solution is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in Southern and Northern blotting, dot blotting, and colony and plaque lifts. 

Solubility
Soluble in water.

Notes
0.3M sodium citrate, 3.0M sodium chloride, pH 7.0.
RUO – Research Use Only

General References:

  1. CJ Quaife.; SD Findley.; JC Erickson.; GJ Froelick. Induction of a new metallothionein isoform (MT-IV) occurs during differentiation of stratified squamous epithelia. Biochemistry. 1994, 33,(23), 7250-7259.
  2. M Cappello.; S Keshav.; C Prince.; DP Jewell.; S Gordon. Detection of mRNAs for macrophage products in inflammatory bowel disease by in situ hybridisation. Gut . 1992, 33,(9), 1214-1219.