Water, RNase, DNase-free
Water, RNase, DNase-free
Water, RNase, DNase-free
Thermo Scientific Chemicals

Water, RNase, DNase-free

CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Catalog NumberQuantity
J60610.EQC
also known as J60610-=C
3.8 L
Catalog number J60610.EQC
also known as J60610-=C
Price (TWD)
-
Quantity:
3.8 L
Request bulk or custom format
Chemical Identifiers
CAS7732-18-5
IUPAC Namewater
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Protease activityNone detected
Appearance (Color)Clear colorless
Assay (special)Bioburden: No growth
RNaseNone detected
View more
Water, RNase, DNase-free high quality, RNase/DNase-free water is used in all molecular biology applications like PCR, DNA. Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and is suitable for all RNA work.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Water, RNase, DNase-free high quality, RNase/DNase-free water is used in all molecular biology applications like PCR, DNA. Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and i
RUO – Research Use Only

General References:

  1. K Aoyagi.; T Tatsuta, M Nishigaki.; S Akimoto. A faithful method for PCR-mediated global mRNA amplification and its integration into microarray analysis on laser-captured cells. Biochemical and Biophysical Research Communications. 2003, 300 ,(4), 915-920.
  2. NR Deleault, RW Lucassen, S Supattapone. RNA molecules stimulate prion protein conversion. Nature. 2003, 425 , 717-720 .