Thermo Scientific Chemicals

Zolmitriptan

Catalog number: J60616.03
1 g, Each
Thermo Scientific Chemicals

Zolmitriptan

Catalog number: J60616.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60616.03
also known as J60616-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
139264-17-8
Chloride content
0.1% max.
Loss on Drying
1.0% max.
Residue on ignition
0.1% max.
Assay (unspecified)
99.0% min.
Assay (unspecified)
Specific rotation: -4.5 to -8.5?

Description

A serotonin 5HT1D-receptor agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text