Thermo Scientific Chemicals

TRIS, 1.0M buffer soln., pH 7.5

Catalog number: J60636.K2
1 L, Each
Thermo Scientific Chemicals

TRIS, 1.0M buffer soln., pH 7.5

Catalog number: J60636.K2
1 L, Each
Quantity
Catalog number: J60636.K2
Price (USD)
Price: 97.40
Online price: 82.65
Your price:
Quantity
-

Description

TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.

Solubility
Soluble in water and organic solvents.

Notes
Store in cool place. Keep container
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text