Thermo Scientific Chemicals

Dihydroergocristine methanesulfonate, Thermo Scientific Chemicals

Catalog number: J60638.03
1 g, Each
Thermo Scientific Chemicals

Dihydroergocristine methanesulfonate, Thermo Scientific Chemicals

Catalog number: J60638.03
1 g, Each
Quantity
Catalog number: J60638.03
also known as J60638-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
24730-10-7
IUPAC Name
(2R,4R,7R)-N-[(1S,2S,4R,7S)-7-benzyl-2-hydroxy-5,8-dioxo-4-(propan-2-yl)-3-oxa-6,9-diazatricyclo[7.3.0.0²,⁶]dodecan-4-yl]-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene-4-carboxamide; methanesulfonic acid
Molecular Formula
C36H45N5O8S
InChI Key
SPXACGZWWVIDGR-SPZWACKZSA-N
SMILES
CS(O)(=O)=O.CC(C)[C@@]1(NC(=O)[C@@H]2C[C@H]3[C@@H](CC4=CNC5=CC=CC3=C45)N(C)C2)O[C@@]2(O)[C@@H]3CCCN3C(=O)[C@H](CC3=CC=CC=C3)N2C1=O
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
≥98.0%

Description

5-HT receptor antagonist; partial agonist at adrenergic and dopaminergic receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text