Thermo Scientific Chemicals

Fasudil monohydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: J60751.03
1 g, Each
Thermo Scientific Chemicals

Fasudil monohydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: J60751.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60751.03
also known as J60751-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
105628-07-7
IUPAC Name
hydrogen 5-(1,4-diazepane-1-sulfonyl)isoquinoline chloride
Molecular Formula
C14H18ClN3O2S
InChI Key
LFVPBERIVUNMGV-UHFFFAOYSA-N
SMILES
[H+].[Cl-].O=S(=O)(N1CCCNCC1)C1=C2C=CN=CC2=CC=C1
Elemental Analysis
Report Results
Purity
≥99.0% (TLC)
Appearance (Color)
White to off-white
Form
Powder or crystals or crystalline powder
Purity
≥99.0% (HPLC)

Description

Fasudil monohydrochloride is used in the prevention and treating cardiovascular and cerebrovascular diseases. It is also used in the inhibition of proliferation of vascular smooth muscle cells. It serves as a selective Rho-associated protein kinase (ROCK) inhibitor. Further, it protects against methyl mercury-induced axonal degeneration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and dimethyl sulfoxide. Insoluble in ethanol.

Notes
Incompatible with acids, bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text