MES, 1.0M buffer soln., pH 6.0
MES, 1.0M buffer soln., pH 6.0
MES, 1.0M buffer soln., pH 6.0
Thermo Scientific Chemicals

MES, 1.0M buffer soln., pH 6.0

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J60763.AP
also known as J60763-AP
Price (TWD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS145224-94-8
IUPAC Name4-(2-sulfonatoethyl)morpholin-4-ium
Molecular FormulaC6H13NO4S
InChI KeySXGZJKUKBWWHRA-UHFFFAOYSA-N
SMILES[O-]S(=O)(=O)CC[NH+]1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless liquid
FormLiquid
CommentComponent is dissolved in UV treated 18.2 megohm-cm ± 1 water, followed by pH adjustment and sub-micron filter.
pH6.0 ± 0.2
MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. d'Errico, C.; Jorgensen, J. O.; Krogh, K. B.; Spodsberg, N.; Madsen, R.; Monrad, R. N. Enzymatic degradation of lignin-carbohydrate complexes (LCCs): Model studies using a fungal glucuronoyl esterase from Cerrena unicolor. Biotechnol. Bioeng. 2015, 112, (5), 914-922.
  2. Yan, Y.; Sun, J.; Zhang, K.; Zhu, H.; Yu, H.; Sun, M.; Huang, D.; Wang, S. Visualizing Gaseous Nitrogen Dioxide by Ratiometric Fluorescence of Carbon Nanodots-Quantum Dots Hybrid. Anal. Chem. 2015, 87 (4), 2087-2093.