Phosphate, 0.5M buffer soln., pH 7.4
Phosphate, 0.5M buffer soln., pH 7.4
Phosphate, 0.5M buffer soln., pH 7.4
Thermo Scientific Chemicals

Phosphate, 0.5M buffer soln., pH 7.4

Buffer solution commonly used in biological research. A high buffering capacity reagent used in molecular biology, biochemistry, and chromatography. | CAS: 7558-80-7119.976 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J60785.AK
also known as J60785-AK
Price (EUR)
83,30
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
83,30
Each
Specifications
CAS7558-80-7
PackagingPlastic Bottle
Recommended StorageAmbient Temperatures
Concentration0.5M buffer soln.
Molecular Weight (g/mol)119.976
View more
Phosphate-buffer (abbreviated PB) is a buffer solution commonly used in biological research. A high buffering capacity reagent used in molecular biology, biochemistry, and chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only