Angiotensin II (human)
Angiotensin II (human)
Angiotensin II (human)
Thermo Scientific Chemicals

Angiotensin II (human)

CAS: 4474-91-31046.197 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 mg
25 mg
Número de catálogo J60866.LB0
también denominado J60866-#0
Precio (USD)
-
Cantidad:
5 mg
Pedido a granel o personalizado
Identificadores químicos
CAS4474-91-3
IUPAC Name(3S)-3-amino-3-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(2S)-1-{[(2S)-1-[(2S)-2-{[(1S)-1-carboxy-2-phenylethyl]carbamoyl}pyrrolidin-1-yl]-3-(1H-imidazol-5-yl)-1-oxopropan-2-yl]carbamoyl}-2-methylbutyl]carbamoyl}-2-(4-hydroxyphenyl)ethyl]carbamoyl}-2-methylpropyl]carbamoyl}-4-[(diaminomethylidene)amino]butyl]carbamoyl}propanoic acid
Molecular FormulaC50H71N13O12
InChI KeyCZGUSIXMZVURDU-GYGDIBSWSA-N
SMILESCC[C@H](C)C(NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)[C@@H](NC(=O)[C@H](CCCN=C(N)N)NC(=O)[C@@H](N)CC(O)=O)C(C)C)C(=O)N[C@@H](CC1=CN=CN1)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Mass SpecConforms
FormPowder
Appearance (Color)White
Assay from Supplier's CofA≥98.0%
Ang II stimulates angiogenesis and increases microvessel density. In addition, the renin-angiotensin and nitric oxide-generating systems appear to interract in the regulation of cardiovascular function. Angiotensin (Ang) II is a peptide which plays an important role in the regulation of blood pressure and has strong vasoconstrictive effects. It also stimulates the release of aldosterone from the adrenal gland and inhibits adenylate cyclase activity. It also increases the entry of Ca2+ in heart muscle via voltage-sensitive channels and activates myosin light chain kinase (MYLK). It also elicits an increase in voltage-dependent delayed rectifier K+ current in neurons and activates JAK2 in smooth muscle cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ang II stimulates angiogenesis and increases microvessel density. In addition, the renin-angiotensin and nitric oxide-generating systems appear to interract in the regulation of cardiovascular function. Angiotensin (Ang) II is a peptide which plays an important role in the regulation of blood pressure and has strong vasoconstrictive effects. It also stimulates the release of aldosterone from the adrenal gland and inhibits adenylate cyclase activity. It also increases the entry of Ca2+ in heart muscle via voltage-sensitive channels and activates myosin light chain kinase (MYLK). It also elicits an increase in voltage-dependent delayed rectifier K+ current in neurons and activates JAK2 in smooth muscle cells.

Solubility
Soluble in Water (25 mg/ml), and 5% Acetic Acid (1 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. keep away from Strong acids, Strong bases .
RUO – Research Use Only

General References:

  1. Caroline Cheng et. Al. Angiotensin II (ATII)-mediated hypertension increases the risk for acute coronary events, which may be caused by augmented collagen degradation. Interstitial fibers of collagen type I in the plaque can be degraded by MMP8 and MMP13 specifically. Atherosclerosis . 2009, 204,(1), 26-33.
  2. Hsiang-Hao Hsu et. Al. Mechanisms of angiotensin II signaling on cytoskeleton of podocytes. Journal of Molecular Medicine . 2008, 86,(12), 1379-1394.
Contenido y almacenamiento
Store at -20°C