Dacarbazine
Dacarbazine
Dacarbazine
Thermo Scientific Chemicals

Dacarbazine

CAS: 4342-03-4 | C6H10N6O | 182.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61023.03
also known as J61023-03
Price (EUR)
239,00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
239,00
Each
Add to cart
Specifications
CAS3-4-4342
Chemical Name or MaterialDacarbazine
ColorWhite to Yellow
EINECS Number224-396-1
Formula Weight182.18
View more
Used in treatment of malignant melanoma and sarcomas. Generates methyl adducts to purine bases in DNA

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used in treatment of malignant melanoma and sarcomas
  • Generates methyl adducts to purine bases in DNA
RUO – Research Use Only