Magnesium sulfate, 1M aq. soln., sterile-filtered
Magnesium sulfate, 1M aq. soln., sterile-filtered
Magnesium sulfate, 1M aq. soln., sterile-filtered
Thermo Scientific Chemicals

Magnesium sulfate, 1M aq. soln., sterile-filtered

CAS: 7487-88-9 | MgO4S | 120.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
125 mL
250 mL
Catalog number J61030.EQE
also known as J61030-=E
Price (USD)
-
Quantity:
125 mL
Request bulk or custom format
Specifications
CAS7487-88-9
ChEBICHEBI:32599
Chemical Name or MaterialMagnesium sulfate
Concentration or Composition (by Analyte or Components)1M aq. soln.
Formula Weight120.37
View more
Magnesium sulfate is applied in replacement therapy for hypomagnesemia. It can be used orally as a laxative to relieve occasional constipation, and to treat low levels of magnesium. It works by replacing magnesium in the body and increasing water in the intestines. It is the first-line antiarrhythmic agent for torsades de pointes in cardiac arrest under the 2005 ECC guidelines and for managing quinidine-induced arrhythmias. It is effective in decreasing the risk that pre-eclampsia progresses to eclampsia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Fully miscible in water. Soluble in alcohol, ether (11.6 mg/ml at 18°C) and glycerin. Insoluble in acetone.

Notes
Stable under recommended storage conditions. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Andrea G.Witlina, (DO); Baha M.Sibai, MD. Magnesium sulfate therapy in preeclampsia and eclampsia. Obstetrics & Gynecology. 1998, 92,(5), 883-889.
  2. Scott W.Snyder, MD; Michael S.Cardwell, MD. Neuromuscular blockade with magnesium sulfate and nifedipine. American Journal of Obstetrics and Gynecology. 1989, 161,(1), 35-36.