Leupeptin Hemisulfate
Leupeptin Hemisulfate
Leupeptin Hemisulfate
Thermo Scientific Chemicals

Leupeptin Hemisulfate

Reversible protease inhibitor which inhibits cathepsin B, calpain and trypsin | CAS: 103476-89-7 | C20H39N6O4+
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number J61188.MC
also known as J61188-MC
Price (TWD)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS103476-89-7
IUPAC Name[amino({[(4S)-4-[(2S)-2-[(2S)-2-acetamido-4-methylpentanamido]-4-methylpentanamido]-5-oxopentyl]amino})methylidene]azanium
Molecular FormulaC20H39N6O4
InChI KeyGDBQQVLCIARPGH-ULQDDVLXSA-O
SMILESCC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=[NH2+])C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (HPLC)≥96.5%
Assay (special)Peptide content: ≥85.0%
Reversible protease inhibitor, which inhibits cathepsin B, calpain and trypsinPharmaceutical composition containing leupeptin hemisulfate is used as an anti-malarial agent. It is employed in the treatment of noise-induced hearing loss. It also protects the heart from myocardial stunning. Further, it is used to inhibit serine, cysteine proteases, plasmin, trypsin, papain, kallikrein and cathepsin B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reversible protease inhibitor, which inhibits cathepsin B, calpain and trypsinPharmaceutical composition containing leupeptin hemisulfate is used as an anti-malarial agent. It is employed in the treatment of noise-induced hearing loss. It also protects the heart from myocardial stunning. Further, it is used to inhibit serine, cysteine proteases, plasmin, trypsin, papain, kallikrein and cathepsin B.

Solubility
Soluble in water, ethanol, dimethyl formamide and acetic acid.

Notes
Store in a cool place. Incompatible with strong acids and strong bases.
RUO – Research Use Only

General References:

  1. Uchikawa, S.; Yoda, M.; Tohmonda, T.; Kanaji, A.; Matsumoto, M.; Toyama, Y.; Horiuchi, K. ADAM17 regulates IL-1 signaling by selectively releasing IL-1 receptor type 2 from the cell surface. Cytokine 2015, 71 (2), 238-245.
  2. Solter, P. F.; Oyama, M. A.; Machen, M. C.; Trafny, D. J.; Sisson, D. D. Detection by ELISA of C-terminal proBNP in plasma from cats with cardiomyopathy. Vet. J. 2015, 206 (2), 213-217.
Contents & Storage
-30°C to -10°C