Citrate, 0.5M buffer soln., pH 4.0
Citrate, 0.5M buffer soln., pH 4.0
Citrate, 0.5M buffer soln., pH 4.0
Thermo Scientific Chemicals

Citrate, 0.5M buffer soln., pH 4.0

CAS: 6132-04-3294.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J61249.AK
Price (EUR)
86,65
Online Exclusive
95,60
Save 8,95 (9%)
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
86,65
Online Exclusive
95,60
Save 8,95 (9%)
Each
Specifications
CAS4-3-6132
ColorColorless
OdorOdorless
Recommended StorageKeep cold
Concentration0.5M buffer soln.
View more
Citrate, 0.5M buffer soln., pH 4.0 may be used for antigen retrieval of tissue samples. The citrate solution is designed to break protein cross-links; thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies. Citrate has anticoagulant activity as a calcium chelator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citrate, 0.5M buffer soln., pH 4.0 may be used for antigen retrieval of tissue samples. The citrate solution is designed to break protein cross-links; thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies. Citrate has anticoagulant activity as a calcium chelator.

Solubility
It is soluble in water.

Notes
Store at room temperature.
RUO – Research Use Only

General References:

  1. JM Chirgwin.; AE Przybyla.; RJ MacDonald. Isolation of biologically active ribonucleic acid from sources enriched in ribonuclease. Biochemistry. 1979, 18,(24), 5294-5299.
  2. PJ Elving.; JM Markowitz.; I Rosenthal. Preparation of buffer systems of constant ionic strength. Analytical Chemistry. 1956, 28,(7), 1179-1180.